Fisher Chemical Glycerol, Certified ACS Grade, 1 L

Representative image only.

Fisher Chemical Glycerol, Certified ACS Grade, 1 L

Fisher Chemical | Mfr: G331| Item: UX-88010-74

Quality and consistency your lab can count on

$421.25USD / Each

$421.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.5%
  • Color (APHA)—≤10
  • Residue after Ignition—≤0.005%
  • Neutrality—To pass test
  • Chlorinated Compounds—≤0.003%
  • Sulfate—≤0.001%
  • Fatty Acid Esters (as Butyric Acid)—≤0.05%
  • Substances Darkened by H2SO4—To pass test
  • Heavy Metals (as Pb)—≤2ppm
  • Water—≤0.5%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaHOCH2CH(OH)CH2OH
  • ChemistrySolvents
  • SynonymsGlycerine
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight92.09
  • PackagingHDPE Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGrey
  • CAS Number56-81-5
  • Boiling Point C (° C)290
  • Melting Point C (° C)18
  • DescriptionGlycerol, Certified ACS Grade, 1 L
Documents & Certificates
Reviews
Q&A