Fisher Chemical Hydrochloric Acid Solution, 1N, Certified Grade, 1 L

Representative image only.

Fisher Chemical Hydrochloric Acid Solution, 1N, Certified Grade, 1 L

Fisher Chemical | Mfr: SA481| Item: EW-88011-88

Quality and consistency your lab can count on

$141.25USD / Each

$141.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Normality — 0.995-1.005
  • Color (APHA)—≤10

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

CorrosiveStorage Code White
Specifications
  • Normality1
  • Purity GradeCertified
  • Concentration1
  • Empirical FormulaHCl
  • MatrixWater
  • ClassReagent Acids and Bases
  • FormLiquid
  • SynonymsMuriatic acid
  • Volume (mL)1000
  • Formula Weight36.45
  • PackagingPoly Bottle
  • DOT InformationHydrochloric Acid; Class 8, Packing Group III
  • Storage SymbolWhite
  • Hazard SymbolCorrosive, Toxic
  • UN NumberUN1789
  • CAS Number7647-01-0
  • Boiling Point C (° C)83
  • Melting Point C (° C)-66
  • DescriptionHydrochloric Acid Solution, 1N, Certified Grade, 1 L
Documents & Certificates
Reviews
Q&A