Fisher Chemical Hydrochloric Acid Solution, 36.5 to 38% (w/w), Certified ACS Plus Grade, Safe-Cote™ Glass Bottle, 500 mL

Representative image only.

Fisher Chemical Hydrochloric Acid Solution, 36.5 to 38% (w/w), Certified ACS Plus Grade, Safe-Cote™ Glass Bottle, 500 mL

Fisher Chemical | Mfr: A144S500| Item: EW-88011-62

Quality and consistency your lab can count on

$210.25USD / Each

$210.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — 36.5-38.0%
  • Color (APHA)—≤10
  • Residue after Ignition—≤4ppm
  • Bromide—≤0.005%
  • Sulfate—≤0.5ppm
  • Sulfite—≤0.8ppmExtractable Organic Substances—To pass test(about 5ppm)Free Chlorine—To pass test(about 0.4ppm)
  • Ammonium—≤3ppm
  • Arsenic and Antimony(as Arsenic)—≤0.005ppm
  • Heavy Metals (as Pb)—≤0.1ppm
  • Phosphate—≤0.05ppm
  • Aluminum—≤0.3ppm
  • Boron—≤0.1ppm
  • Calcium—≤0.3ppm
  • Chromium—≤0.2ppm
  • Copper—≤0.1ppm
  • Gold—≤0.3ppm
  • Iron—≤0.1ppm
  • Lead—≤0.1ppm
  • Magnesium—≤0.3ppm
  • Manganese—≤0.3ppm
  • Nickel—≤0.1ppm
  • Potassium—≤0.3ppm
  • Sodium—≤0.3ppm
  • Tin—≤0.3ppm
  • Titanium—≤0.3ppm
  • Zinc—≤0.3ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

CorrosiveToxicStorage Code White
Specifications
  • Concentration36.5 to 38%
  • Purity GradeCertified ACS Plus
  • MatrixWater
  • Empirical FormulaHCl
  • ClassReagent Acids and Bases
  • SynonymsMuriatic acid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight36.45
  • PackagingSafety Coated Glass Bottle
  • DOT InformationHydrochloric Acid; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive, Toxic
  • CAS Number7647-01-0
  • UN NumberUN1789
  • Boiling Point C (° C)57
  • Melting Point C (° C)-35
  • DescriptionHydrochloric Acid Solution, 36.5 to 38% (w/w), Certified ACS Plus Grade, Safe-Cote™ Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A