Fisher Chemical Isooctane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L

Representative image only.

Fisher Chemical Isooctane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L

Fisher Chemical | Mfr: O296SK4| Item: EW-88013-30

Quality and consistency your lab can count on

$516.00USD / Each

$516.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • Color (APHA)—≤10
  • Optical Absorbance at 205nm—≤1.00
  • at 225nm—≤0.10
  • at 254nm—≤0.014
  • Fluorescence Background (as Quinine Sulfate)—To pass test
  • Refractive Index at 25°C—1.3885-1.3905
  • Residue after Evaporation—≤4ppm
  • Water—≤0.02%
  • Water-soluble Titratable Acid—≤0.0003mEq/g
  • Density at 25°C—≤0.690g/mL
  • Sulfur Compounds (as S)—≤0.005%
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeHPLC
  • Empirical Formula(CH3)3CCH2CH(CH3)2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsOctane, Iso-, 2,2,4-Trimethylpentane
  • Volume (mL)4000
  • Formula Weight114.23
  • PackagingSafe-Cote™ Amber Glass Bottle
  • DOT InformationOctanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • UN NumberUN1262
  • CAS Number540-84-1
  • Boiling Point C (° C)98
  • Melting Point C (° C)-107
  • DescriptionIsooctane, HPLC Grade, Safe-Cote™ Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A