Fisher Chemical Methylene Chloride, GC Resolv™ Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Methylene Chloride, GC Resolv™ Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: D1544| Item: EW-88015-68

Quality and consistency your lab can count on

$507.00USD / Each

$507.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Color (APHA) — ≤5
  • Certified for EPA Method 1625 — To pass test
  • ECD-responsive Substances (as Heptachlor Epoxide)† — 5ppt maximum peak or 50ppt total
  • FID-responsive Substances (as Decane)† — 1ppb maximum peak or 10ppb total
  • Free Halogens — ≤1ppm
  • Preservative (Amylene) — 40-200ppm maximum
  • Residue after Evaporation — ≤1ppm
  • Titratable Acid — ≤0.0002mEq/g
  • Water — ≤0.01%
  • Packaged under Nitrogen.
  • †After retention time of Toluene.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Gray
Specifications
  • Purity GradeGC Resolv
  • Empirical FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDichloromethane, DCM
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingGlass Bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolFlammable, Toxic
  • CAS Number75-09-2
  • UN NumberUN1593
  • Boiling Point C (° C)39
  • Melting Point C (° C)-97
  • DescriptionMethylene Chloride, GC Resolv™ Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A