Fisher Chemical Methylene Chloride, OPTIMA™ Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Methylene Chloride, OPTIMA™ Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: D1514| Item: EW-88016-02

Quality and consistency your lab can count on

$463.75USD / Each

$463.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.9%
  • Color (APHA) — ≤5
  • Certified for EPA Test #1625 — To pass test
  • Pesticide Residue Analysis (as Heptachlor Epoxide) — ≤10ng/L
  • Optical Absorbance at 233nm — ≤1.00
  • at 240nm — ≤0.12
  • at 254nm — ≤0.005
  • Fluorescence Background (as Quinine Sulfate) — ≤1ppb
  • Preservative (Amylene) — 40-200ppm
  • Water — ≤0.01%
  • Residue after Evaporation — ≤1ppm
  • Refractive Index at 25°C — 1.4200-1.4230
  • Density at 25°C — 1.315-1.321g/mL
  • Titratable Acid — ≤0.0002mEq/g
  • Free Halogens — To pass test
  • Packaged under Nitrogen, Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Gray
Specifications
  • Purity GradeOPTIMA
  • Empirical FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDichloromethane, DCM
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingGlass Bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolFlammable, Toxic
  • CAS Number75-09-2
  • UN NumberUN1593
  • Boiling Point C (° C)39
  • Melting Point C (° C)-97
  • DescriptionMethylene Chloride, OPTIMA™ Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A