Fisher Chemical Methylene Chloride, Stabilized, Certified ACS Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Methylene Chloride, Stabilized, Certified ACS Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: D374| Item: EW-88016-30

Quality and consistency your lab can count on

$398.00USD / Each

$398.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ≥99.5%
  • Color (APHA) — ≤10
  • Density at 25°C — 1.315-1.321g/mL
  • Residue after Evaporation — ≤0.002%
  • Titratable Acid — ≤0.0003mEq/g
  • Water — ≤0.02%
  • Free Halogens — To pass test

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Gray
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDichloromethane, DCM
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingGlass Bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolFlammable, Toxic
  • CAS Number75-09-2
  • UN NumberUN1593
  • Boiling Point C (° C)39
  • Melting Point C (° C)-97
  • DescriptionMethylene Chloride, Stabilized, Certified ACS Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A