Fisher Chemical Methylene Chloride, Stabilized, HPLC Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Methylene Chloride, Stabilized, HPLC Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: D1434| Item: EW-88016-14

Quality and consistency your lab can count on

$280.50USD / Each

$280.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (including preservative) — ¡Ý99.9%
  • Color (APHA) — ¡Ü10
  • Optical Absorbance at 235nm — ¡Ü1.00
  • at 240nm — ¡Ü0.20
  • at 254nm — ¡Ü0.014
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25¡ãC — 1.4200-1.4230
  • Free Halogens — To pass test
  • Residue after Evaporation — ¡Ü2ppm
  • Preservative:
  • Amylene — 40-200ppm
  • or Cyclohexene — 40-100ppm
  • Water — ¡Ü0.02%
  • Titratable Acid — ¡Ü0.0003mEq/g
  • Density at 25¡ãC — 1.315-1.321g/mL
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Gray
Specifications
  • Purity GradeHPLC
  • Empirical FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsDichloromethane, DCM
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight84.93
  • PackagingGlass Bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolFlammable, Toxic
  • CAS Number75-09-2
  • UN NumberUN1593
  • Boiling Point C (° C)39
  • Melting Point C (° C)-97
  • DescriptionMethylene Chloride, Stabilized, HPLC Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A