Fisher Chemical Petroleum Ether, Certified ACS Grade, 20 L

Representative image only.

Fisher Chemical Petroleum Ether, Certified ACS Grade, 20 L

Fisher Chemical | Mfr: E13920| Item: EW-88072-22

Quality and consistency your lab can count on

$828.00USD / Each

$828.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeCertified ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsLigroine, Benzine, Naphtha Petroleum, Naphtha Solvent
  • Volume (mL)20000
  • PackagingSteel Pail
  • DOT InformationPetroleum Distillates, N.O.S.; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number8032-32-4
  • UN NumberUN1268
  • Boiling Point C (° C)38
  • DescriptionPetroleum Ether, Certified ACS Grade, 20 L
Documents & Certificates
Reviews
Q&A