Fisher Chemical Petroleum Ether, Certified ACS Grade, 500 mL

Representative image only.

Fisher Chemical Petroleum Ether, Certified ACS Grade, 500 mL

Fisher Chemical | Mfr: E139500| Item: EW-88019-10

Quality and consistency your lab can count on

$90.90USD / Each

$90.90USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤10
  • Boiling Range — 36°-60°C
  • Initial Boiling Temperature — 36°-39°C
  • Dry Flask End Point Temperature — 52°-60°C
  • Fraction Distills under 54°C — ≥95%
  • Fraction Distills under 40°C — ≥60%
  • Distillation Residue — To pass test
  • Residue after Evaporation — ≤0.001%
  • Acidity — To pass test
  • Heavy Oils and Fats — To pass test
  • Doctor Test — Sweet
  • Copper Strip Corrosion Test — To pass test
  • Stearin Evaporation Residue — ≤0.003g
  • Saponification Value — ≤0.015mg KOH/g
  • Specific Gravity at 15.5°C — 0.630-0.660
  • Unsaturated Compounds — Trace (≤0.5%)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeCertified ACS
  • ClassGeneral Solutions
  • ChemistrySolutions
  • SynonymsLigroine, Benzine, Naphtha Petroleum, Naphtha Solvent
  • FormLiquid
  • Volume (mL)500
  • PackagingGlass Bottle
  • DOT InformationPetroleum Distillates, N.O.S.; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number8032-32-4
  • UN NumberUN1268
  • Boiling Point C (° C)38
  • DescriptionPetroleum Ether, Certified ACS Grade, 500 mL
Documents & Certificates
Reviews
Q&A