Fisher Chemical Petroleum Ether, Optima™ Grade, 4 L

Representative image only.

Fisher Chemical Petroleum Ether, Optima™ Grade, 4 L

Fisher Chemical | Mfr: E1204| Item: EW-88019-04

Quality and consistency your lab can count on

$403.75USD / Each

$403.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Color (APHA) — ≤5
  • Acidity — To pass test
  • Pesticide Residue Analysis (as Heptachlor Epoxide) — ≤10ng/L
  • Water — ≤0.01%
  • Residue after Evaporation — ≤1ppm
  • Boiling Range (1 drop to dryness) — 35°-60°C
  • Refractive Index at 25°C — ≤1.3560-1.3660
  • Packaged under Nitrogen, Submicron Filtered.
  • Keep tightly closed in a cool place.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsLigroine, Benzine, Naphtha Petroleum, Naphtha Solvent
  • Volume (mL)4000
  • PackagingAmber Glass Bottle
  • DOT InformationPetroleum Distillates, N.O.S.; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number8032-32-4
  • UN NumberUN1268
  • Boiling Point C (° C)38
  • DescriptionPetroleum Ether, Optima™ Grade, 4 L
Documents & Certificates
Reviews
Q&A