Fisher Chemical Potassium Hydroxide Solution, 0.1N in Isopropanol, Certified Grade, 500 mL

Representative image only.

Fisher Chemical Potassium Hydroxide Solution, 0.1N in Isopropanol, Certified Grade, 500 mL

Fisher Chemical | Mfr: ST110500| Item: EW-88085-61

Quality and consistency your lab can count on

$185.50USD / Each

$185.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableStorage Code Red
Specifications
  • Normality0.1
  • Concentration0.1N
  • Purity GradeCertified
  • Empirical FormulaKOH
  • MatrixIsopropanol
  • ClassReagent Acids and Bases
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • Formula Weight56.11
  • PackagingHDPE Bottle
  • DOT InformationIsopropyl alcohol, Potassium hydroxide; Class 3 (8), Packing Group II
  • Storage SymbolRed
  • Hazard SymbolCorrosive, Flammable
  • UN NumberUN2924
  • CAS Number1310-58-3
  • Boiling Point C (° C)82.8
  • Melting Point C (° C)-88.9
  • DescriptionPotassium Hydroxide Solution, 0.1N in Isopropanol, Certified Grade, 500 mL
Documents & Certificates
Reviews
Q&A