Fisher Chemical Sodium Acetate Anhydrous, Certified ACS Grade, 500 g

Representative image only.

Fisher Chemical Sodium Acetate Anhydrous, Certified ACS Grade, 500 g

Fisher Chemical | Mfr: S210500| Item: UX-88022-70

Quality and consistency your lab can count on

$353.00USD / Each

$353.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • pH of a 5% Solution at 25°C — 7.0 to 9.2
  • Chloride — ≤0.002%
  • Phosphate — ≤0.001%
  • Sulfate — ≤0.003%
  • Calcium — ≤0.005%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.001%
  • Loss on Drying at 120°C — ≤1.0%
  • Insoluble Matter — ≤0.01%
  • Magnesium — ≤0.002%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaC2H3O2Na
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight82.03
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number127-09-3
  • Melting Point C (° C)324
  • DescriptionSodium Acetate Anhydrous, Certified ACS Grade, 500 g
Documents & Certificates
Reviews
Q&A