Fisher Chemical Sodium Chloride, BP/EP/FCC/JP/USP Grade, 500 g

Representative image only.

Fisher Chemical Sodium Chloride, BP/EP/FCC/JP/USP Grade, 500 g

Fisher Chemical | Mfr: S642500| Item: UX-88023-56

Quality and consistency your lab can count on

$191.50USD / Each

$191.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity or Alkalinity — To pass test
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Magnesium and Alkaline Earth Metals (as Ca) — ≤0.01%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iodides — To pass test
  • Bromides — ≤0.01%
  • Iron — ≤2ppm
  • Loss on Drying at 105°C — To pass test
  • Ferrocyanide — To pass test
  • Sulfate — ≤0.020%
  • Aluminum — ≤0.2ppm
  • Nitrites — To pass test
  • Limit of Phosphates — ≤0.0025%
  • Meets FCC Chemical Specifications
  • Assay (after drying at 625°C) — 97.5-100.5%
  • Arsenic — ≤1mg/kg
  • Calcium and Magnesium — ≤2.0%
  • Heavy Metals (as Pb) — ≤2mg/kg
  • Loss on Drying at 110°C — ≤0.5%
  • Meets EP and BP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity and Alkalinity — To pass test
  • Bromides — ≤50ppm
  • Ferrocyanides — To pass test
  • Iodides — To pass test
  • Nitrites — To pass test
  • Phosphates — ≤25ppm
  • Sulfates — ≤200ppm (EP); ≤250ppm (BP)
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Heavy Metals (as Pb) — ≤5ppm (EP); ≤10ppm (BP)
  • Iron — ≤2ppm (EP); ≤20ppm (BP)
  • Magnesium and A

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeBP/EP/FCC/JP/USP
  • Empirical FormulaNaCl
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • SynonymsHalite, Common salt, Rock salt
  • FormSolid
  • Weight (g)500
  • Formula Weight58.44
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7647-14-5
  • Boiling Point C (° C)1461
  • Melting Point C (° C)801
  • DescriptionSodium Chloride, BP/EP/FCC/JP/USP Grade, 500 g
Documents & Certificates
Reviews
Q&A