Fisher Chemical™ Sodium Chloride

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$156.00 - $1,972.00USD / Each

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Weight (g) Description
Availability Pricing
Fisher Chemical Sodium Chloride, Certified Grade, 500 g EW-88023-44 Mfr # S671500
COMPARE
Certified 500 Sodium Chloride, Certified Grade, 500 g
$156.00USD / Each
Fisher Chemical Sodium Chloride, Certified ACS Grade, 1 kg EW-88023-52 Mfr # S2711
COMPARE
Certified ACS 1000 Sodium Chloride, Certified ACS Grade, 1 kg
$180.00USD / Each
Fisher Chemical Sodium Chloride, Certified ACS Grade, 3 kg EW-88023-54 Mfr # S2713
COMPARE
Certified ACS 3000 Sodium Chloride, Certified ACS Grade, 3 kg
$382.25USD / Each
Fisher Chemical Sodium Chloride, BP/EP/FCC/JP/USP Grade, 500 g EW-88023-56 Mfr # S642500
COMPARE
BP/EP/FCC/JP/USP 500 Sodium Chloride, BP/EP/FCC/JP/USP Grade, 500 g
$191.50USD / Each
Fisher Chemical Sodium Chloride, Certified ACS Grade, 10 kg EW-88080-54 Mfr # S27110
COMPARE
Certified ACS 10000 Sodium Chloride, Certified ACS Grade, 10 kg
$493.75USD / Each
Fisher Chemical Sodium Chloride, Certified ACS Grade, 350 lg EW-88080-55 Mfr # S271350LB
COMPARE
Certified ACS 158757 Sodium Chloride, Certified ACS Grade, 350 lg
$1,972.00USD / Each
Fisher Chemical Sodium Chloride, Certified ACS Grade, 50 kg EW-88080-56 Mfr # S27150
COMPARE
Certified ACS 50000 Sodium Chloride, Certified ACS Grade, 50 kg
$1,038.00USD / Each
Fisher Chemical Sodium Chloride, FCC/USP Grade, 10 kg EW-88082-84 Mfr # S64010
COMPARE
FCC/USP 10000 Sodium Chloride, FCC/USP Grade, 10 kg
$391.25USD / Each
Fisher Chemical Sodium Chloride, FCC/USP Grade, 3 kg EW-88082-85 Mfr # S6403
COMPARE
FCC/USP 3000 Sodium Chloride, FCC/USP Grade, 3 kg
$336.50USD / Each
Fisher Chemical Sodium Chloride, FCC/USP Grade, 500 g EW-88082-88 Mfr # S640500
COMPARE
FCC/USP 500 Sodium Chloride, FCC/USP Grade, 500 g
$188.00USD / Each
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat