Fisher Chemical Sodium Chloride, Certified Grade, 500 g

Representative image only.

Fisher Chemical Sodium Chloride, Certified Grade, 500 g

Fisher Chemical | Mfr: S671500| Item: EW-88023-44

Quality and consistency your lab can count on

$156.00USD / Each

$156.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Insoluble Matter — ≤0.005%
  • Free Alkali (as NaOH) — None
  • Iodide — ≤0.002%
  • Phosphate — ≤5ppm
  • Barium — To pass test
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤2ppm
  • Chlorate — ≤0.001%
  • Free Acid (as HCl) — ≤0.002%
  • Bromide — ≤0.01%
  • Nitrogen Compounds (as N) — ≤0.001%
  • Sulfate — ≤0.004%
  • Calcium, Magnesium and R2O3 Precipitate — ≤0.005%
  • Potassium — ≤0.005%
  • Nitrate — ≤0.003%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeCertified
  • Empirical FormulaNaCl
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • SynonymsHalite, Common salt, Rock salt
  • FormSolid
  • Weight (g)500
  • Formula Weight58.44
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7647-14-5
  • Boiling Point C (° C)1461
  • Melting Point C (° C)801
  • DescriptionSodium Chloride, Certified Grade, 500 g
Documents & Certificates
Reviews
Q&A