Fisher Chemical Sodium Chloride, Certified ACS Grade, 10 kg

Representative image only.

Fisher Chemical Sodium Chloride, Certified ACS Grade, 10 kg

Fisher Chemical | Mfr: S27110| Item: UX-88080-54

Quality and consistency your lab can count on

$493.75USD / Each

$493.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeCertified ACS
  • Empirical FormulaNaCl
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsHalite, Common salt, Rock salt
  • Weight (g)10000
  • Formula Weight58.44
  • PackagingPoly Pail
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7647-14-5
  • Boiling Point C (° C)1461
  • Melting Point C (° C)801
  • DescriptionSodium Chloride, Certified ACS Grade, 10 kg
Documents & Certificates
Reviews
Q&A