Fisher Chemical Toluene, HPLC Grade, 4 L

Representative image only.

Fisher Chemical Toluene, HPLC Grade, 4 L

Fisher Chemical | Mfr: T2904| Item: EW-88041-08

Quality and consistency your lab can count on

$358.00USD / Each

$358.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.8%
  • Color (APHA)—≤10
  • Optical Absorbance at 286nm—≤1.00
  • Refractive Index at 25°C—1.4930-1.4950
  • Residue after Evaporation—≤10ppm
  • Substances Darkened by H2SO4—To pass test
  • Water—≤0.02%
  • Sulfur Compounds (as S)—≤0.003%
  • Submicron filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeHPLC
  • Empirical FormulaC6H5CH3
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsMethylbenzene, Toluol, Phenyl Methane
  • Volume (mL)4000
  • Formula Weight92.14
  • PackagingGlass Bottle
  • DOT InformationToluene; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Harmful
  • CAS Number108-88-3
  • UN NumberUN1294
  • Boiling Point C (° C)111
  • Melting Point C (° C)-95
  • DescriptionToluene, HPLC Grade, 4 L
Documents & Certificates
Reviews
Q&A