Fisher Chemical Tris Base, Molecular Biology Grade, 500 g

Representative image only.

Fisher Chemical Tris Base, Molecular Biology Grade, 500 g

Fisher Chemical | Mfr: BP152500| Item: EW-88041-97

Quality and consistency your lab can count on

$319.50USD / Each

$319.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (corrected for H2O) — ≥99.8%
  • Absorbance of a 1M Solution at 280nm—≤0.05
  • Water—≤2%
  • pH of a 0.05M Solution at 25°C—10.3 to 10.5
  • Copper—≤1ppm
  • Iron—≤2ppm
  • Magnesium—≤10ppm
  • Lead—≤1ppm
  • Arsenic—≤5ppm
  • Electrophoresis—To pass test
  • DNase—Not detected
  • RNase—Not detected
  • Protease—Not detected

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeMolecular Biology
  • Empirical FormulaH2NC(CH2OH)3
  • ClassBuffers
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsTrimethylol Aminomethane, Tris Buffer, Tris(hydroxymethyl)aminomethane, Trizma, THAM
  • Weight (g)500
  • Formula Weight121.14
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolFlammable, Irritant
  • CAS Number77-86-1
  • Boiling Point C (° C)219
  • Melting Point C (° C)168.5
  • DescriptionTris Base, Molecular Biology Grade, 500 g
Documents & Certificates
Reviews
Q&A