Fisher Chemical Xylenes, Certified ACS Grade, 4 L

Representative image only.

Fisher Chemical Xylenes, Certified ACS Grade, 4 L

Fisher Chemical | Mfr: X54| Item: EW-88043-24

Quality and consistency your lab can count on

$539.00USD / Each

$539.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (Xylene isomers plus Ethylbenzene [Ethylbenzene not to exceed 25%]) — ≥98.5%
  • Color (APHA)—≤10
  • Residue after Evaporation—≤0.002%
  • Sulfur Compounds (as S)—≤0.003%
  • Substances Darkened by H2SO4—To pass test
  • Water—≤0.05%
  • Heavy Metals (as Pb)—≤0.1ppm
  • Copper—≤0.1ppm
  • Iron—≤0.1ppm
  • Nickel—≤0.1ppm
  • Silver—≤0.1ppm

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Red
Specifications
  • Concentration0.96
  • Purity GradeCertified ACS
  • Empirical FormulaC6H4(CH3)2
  • MatrixEthylbenzene
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsXylol, Dimethylbenzene
  • Volume (mL)4000
  • Formula Weight106.17
  • PackagingGlass Bottle
  • DOT InformationXylenes; Class 3, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolHarmful
  • CAS Number1330-20-7
  • UN NumberUN1307
  • Boiling Point C (° C)136
  • Melting Point C (° C)-34
  • DescriptionXylenes, Certified ACS Grade, 4 L
Documents & Certificates
Reviews
Q&A