Fluke 377 FC Noncontact Voltage True-rms Clamp Meter with iFlex® Probe

Representative image only.

Fluke 377 FC Noncontact Voltage True-rms Clamp Meter with iFlex® Probe

Fluke | Mfr: Fluke-377 FC| Item: EW-20051-14

FieldSense™ technology allows faster and safer voltage and current testing without contact

$763.20USD / Each
$848.00USD / Each

$763.20USD / Each
$848.00USD / Each

Buy It Together

Current Item

Fluke 377 FC Noncontact Voltage True-rms Clamp Meter with iFlex® Probe
Fluke 377 FC Noncontact Voltage True-rms Clamp Meter with iFlex® Probe
2005114

$763.20USD / Each
$848.00USD / Each
+
Fluke TL224 Flexible Silicone Test Lead Set
2600225

$42.30USD / Set of 2
$47.00USD / Set of 2
=
Total Price 805.50 ADD 2 ITEMS TO CART
Product Description

Includes

iFlex flexible current probe (48 cm; 18"), TL224 silicone insulated test leads, TP175 adjustable length test probes, AC285 grounding clip, TPAK magnetic hanging kit, with 23 cm (9") hanging strap, and carrying case.

Key Features

  • Get accurate voltage and current measurements using the clamp jaw
  • Measure extremely high current with iFlex® probe
  • Take three-phase voltage and current tests in three quick steps
  • Color changing screen indicates stable measurements
  • 3 year warranty

More About this Item

The Fluke 377 FC and 378 FC clamp meters use FieldSense technology for safer and faster voltage and current measurements. Simply clip the black test lead to any electrical ground then place the clamp jaw around the conductor. The clamp jaw quickly measures voltage and current without touching any live wires then displays the measurement on the meter. The 34 mm (1-5/16") jaw opening is ideal for use in small compartments or boxes with tightly arranged electrical wires. For your convenience and safety, the display turns green when a stable FieldSense measurement is detected. Use the included 48 cm (18") diameter iFlex probe to measure extremely high current measurements. The probe handles AC current as high as 2500 A and provides access to large conductors in tight spaces.

Meters also completes three-phase measurements in quick, easy steps. A full set of phase-to-ground and phase-to-phase values are calculated and shown on the meter. Use the Fluke Connect® software to view the values as well as auto-calculated phase rotation on your smart phone. The screen turns bright green when continuity is detected—ideal for noisy work areas. The included TPAK magnetic hanging kit comes with a magnet and a hook strap for nonmagnetic surfaces to hang your meter for convenient hands-free operation.

3 Year Warranty
Specifications
  • DC Max (Volts)600
  • DC Voltage Accuracy1% ± 5 digits
  • Current (AC/DC)Jaw: 999.9 A (AC/DC); iFlex probe: 2500 A (AC)
  • AC Max (Volts)1000
  • Ac Voltage AccuracyFieldSense
    ≤ 4/0 AWG: 3% ±5 digits (45 Hz to 66 Hz)
    ≥ 4/0 AWG: 5% ±5 digits (45 Hz to 66 Hz)
    Test leads1% ±5 digits (20 Hz to 500 Hz)
  • AC Current AccuracyJaw: 2% ± 5 digits (45 Hz to 66 Hz); Iflex: 3% ±5 digits (5 Hz to 500 Hz)
  • Resistance Max (Ω)600
  • Resistance Max (KΩ)6
  • Resistance Max (MΩ)0.06
  • Capacitance Max (mF)1
  • Capacitance Accuracy1% ±4 digits
  • Resistivity Accuracy1% ±5 digits
  • ContinuityYes
  • True RmsYes
  • Min Temperature (° F)14
  • Max Temperature (° F)122
  • Min Temperature (° C)-10
  • Max Temperature (° C)50
  • Current FrequencyJaw: 45 to 66 Hz; iflex: 5 to 500 Hz
  • DisplayDigital, dual readout
  • Rf Communications2405 MHz to 2480 MHz, <100 mw output power
  • Safety RatingIEC 61010-1: Pollution degree 2
  • PowerTwo AA (alkaline)
  • Width (in)3.4
  • Length (in)10.8
  • Height (in)1.9
  • Width (cm)8.6
  • Length (cm)27.4
  • Height (cm)4.7
  • DescriptionNoncontact Voltage True-rms Clamp Meter with iFlex® Probe
  • Warranty3 years
Reviews
Q&A