Fluke 393 FC Solar Clamp Meter, IRR1 Irradiance Meter, and MC4 Test Leads Bundle
Representative image only.
Fluke 393 FC Solar Clamp Meter, IRR1 Irradiance Meter, and MC4 Test Leads Bundle
Fluke | Mfr: FLK-393-IRR-PVLEAD| Item: EW-20051-22
Measure safely with this CAT III 1500 V rated clamp meter
$1,304.00USD / Each
sign in for your price.
$1,304.00USD / Each
sign in for your price.
Product Description
Includes
Fluke 393 FC CAT III 1500 V TRMS clamp meter, test leads (CAT III 1500 V rated, right angle plugs, with safety caps), iFlex 18 inch flexible current probe, TPAK magnetic hanging strap, premium carrying case, FLK-IRR1-SOL solar irradiance meter, FLK80PR-IRR external temperature probe with suction cup, C250 carrying case with shoulder strap, four AA alkaline batteries (installed), Pomona PVLEAD1 MC4 to 4 mm test lead set, and Pomona PVLEAD3 MC4 solar clamp test lead set.
Key Features
  • Thin jaw for access to cables in crowded combiner boxes
  • Sturdy IP54 rated for outdoor use
  • DC power measurement, showing readings in kVA
  • Audio Polarity indicator helps prevent accidental miswires
  • Visual Continuity turns provide a bright green light in the display, ideal when working in dark and noisy environments
  • Logging and reporting of test results via Fluke Connect software
  • When measuring AC current the included iFlex flexible current probe gives you unparalleled access to cable is tight spaces
  • Measure solar irradiance, ambient and PV module temperature, array orientation and tilt angles
  • MC4 test lead sets are designed for use with solar clamp meters that accept 4 mm banana plugs, rated to CAT III 1000 V / CAT IV 600 V
  • 3 year warranty
More About this Item
The 393 FC CAT III 1500 V True-rms Clamp Meter with iFlex is designed for technicians who work in high voltage DC environment: solar arrays, wind power, electric railways, data centers battery banks for uninterruptible power supplies. The clamp will measure up to 1500 V DC, 1000 V AC, and up to 999.9 A DC or AC through the clamp jaw. The included iFlex flexible current probe extended AC current measurements up to 2500 amps. This clamp has a thin jaw, giving you access to cables in crowded combiner boxes. Test leads are designed with your work in mind, and are also rated to CAT III 1500 V DC.
Bundles are also available, combining the clamp meter with a wide array of solar tools to easily meet your needs.
3 Year Warranty
Specifications
  • DC Max (Volts)1500
  • DC Voltage Accuracy1 % RD + 5 digits
  • DC Max (amps)999.9
  • DC Current Accuracy2 % RD + 5 digits
  • AC Max (Volts)1000
  • Ac Voltage Accuracy1 % RD + 5 digits (20 Hz to 500 Hz)
  • AC Max (amps)999.9
  • AC Current Accuracy2 % + 5 digits (10 Hz to 100 Hz), 2.5 % + 5 digits (100 Hz to 500 Hz) with jaw3 % RD + 5 digits (10 Hz to 500 Hz) with flexible current probe
  • Resistance Max (KΩ)60
  • Capacitance Max (mF)1
  • Capacitance Accuracy1 % RD + 5 digits
  • True RmsYes
  • DisplayDual display with backlight
  • Power2 AA IEC LR6 alkaline batteries
  • DescriptionSolar Clamp Meter, IRR1 Irradiance Meter, and MC4 Test Leads Bundle
  • Warranty3 Years