Fluke 80T-150UA Type-T Thermocouple Probe
Representative image only.
Fluke 80T-150UA Type-T Thermocouple Probe
Fluke | Mfr: 80T-150UA| Item: EW-26052-60
Offers more flexible strain relief for a long life
$370.79USD / Each
$411.99USD / Each
for an additional
$220.50USD / Each
for an additional
$170.90USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$370.79USD / Each
$411.99USD / Each
Buy It Together
Current Item
Fluke 80T-150UA Type-T Thermocouple Probe
Fluke 80T-150UA Type-T Thermocouple Probe
2605260
$370.79USD / Each
$411.99USD / Each
+ =
Total Price 541.69 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
The innovative SureGrip™ design has a soft rubber handle combined with a ergonomic shape is so comfortable to hold that you’ll forget about the probe and focus on the measurement. All SureGrip probes have an improved, more flexible strain relief for a long life.
1 Year Warranty
Specifications
  • Probe TypeT
  • Min Temperature (° C)-50
  • Max Temperature (° C)150
  • Temperature Accuracy±1°C from 0 to 100°C
  • Connector TypeMini ANSI
  • DescriptionType-T Thermocouple Probe