Fluke 9040 Digital Phase Rotation Indicator
Representative image only.
Fluke 9040 Digital Phase Rotation Indicator
Fluke | Mfr: FLUKE-9040| Item: EW-20021-41
$408.60USD / Each
$454.00USD / Each
$408.60USD / Each
$454.00USD / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
The Fluke 9040 is effective for measuring phase rotation in all areas where three-phase supplies are used to feed motors, drives and electrical systems. This unit is a rotary field indicator and can provide clear indication of the three-phase via an LCD display adn the phase rotation direction to determine correct connections. It allows rapid determination of phase sequence and has a voltage (up to 700V) and frequency range suitable for commercial and industrial applications. The included test probes have a variable clamping range for safe contact, also in industrial sockets.
FEATURES
  • 3 phase indication
  • Indication of phase rotation
  • Clear LCD display
  • No battery required
Two year warranty
Specifications
  • DC Max (Volts)700
  • AC Max (Volts)700
  • Safety RatingCAT III 600V, CAT IV 300V, EN 61010-1
  • DescriptionDigital Phase Rotation Indicator
  • Warranty2 Years