Fluke Calibration 3376880 Bare Thermistor Probe, PTFEinfo-Con
Representative image only.
Fluke Calibration 3376880 Bare Thermistor Probe, PTFEinfo-Con
Fluke Calibration | Mfr: 5611T-P| Item: EW-90450-23
$1,245.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,245.00USD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
5611T Series – PTFE-encapsulated secondary thermistor probes: The 5611T is flexible from one end to the other, and its PTFE coating makes it immersible in acids and other harsh chemicals. This very versatile thermistor has a 3.1 mm (1/8”) tip for measuring in tight places. Its low self-heating also makes it a good choice for measurements in air.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeInfo-Con
  • Sheath Diameter (in)0.125
  • DescriptionBare Thermistor Probe, PTFEinfo-Con