Fluke Calibration 3377310 Immersion Thermistor Probeinfo-Con
Representative image only.
Fluke Calibration 3377310 Immersion Thermistor Probeinfo-Con
Fluke Calibration | Mfr: 5665-P| Item: EW-90451-04
$1,205.00USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,205.00USD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
5665 Series - Miniature immersion secondary thermistor probes: Feature the stability necessary for use as high-accuracy thermometry standards, and if you''re looking for economical lab-grade thermistor probes for accurate work across a narrow temperature range.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeInfo-Con
  • Sheath Diameter (in)0.125
  • DescriptionImmersion Thermistor Probeinfo-Con