Fluke Calibration 9172-INSB Well Insert with Comparison Holes for 9172 Metrology Well, Brass
Representative image only.
Fluke Calibration 9172-INSB Well Insert with Comparison Holes for 9172 Metrology Well, Brass
Fluke Calibration | Mfr: 9172-INSB| Item: EW-90919-44
Ensure accuracy by using the correct size hole to calibrate
$945.00USD / Each
$1,050.00USD / Each
$945.00USD / Each
$1,050.00USD / Each
Product Description
Key Features
  • Three different size holes (3/16", 1/4", and 3/8") to ensure a best fit
  • Stable to ±0.005°C, over their full range
  • Provide radial uniformity of ±0.01 °C
  • Six holes—2 x 3/16", 2 x 1/4", 2 x 3/8"
  • Brass construction
  • 1 year warranty
More About this Item
Hart Scientific is now Fluke Calibration.
1 Year Warranty
Specifications
  • MaterialBrass
  • Compatible WithFluke Calibration 9172 metrology well 90919-10 and -12
  • DescriptionWell Insert with Comparison Holes for 9172 Metrology Well, Brass