Fluke FLK-1AC-A1-II-5PK Pocket-Sized Voltage Detectors, 90 to 1000 V AC; 5/PK
Representative image only.
Fluke FLK-1AC-A1-II-5PK Pocket-Sized Voltage Detectors, 90 to 1000 V AC; 5/PK
Fluke | Mfr: FLK-1AC-A1-II-5PK| Item: EW-20021-36
Detect voltage quickly and easily – without contacting metal!
$180.00USD / Each
$200.00USD / Each
$180.00USD / Each
$200.00USD / Each
Product Description
Includes
2 AAA batteries
Key Features
  • Detects voltage without metal contact
  • Fits in shirt pocket for convenience
  • All outer surfaces are nonconductive for safety
  • 2 year warranty
More About this Item
These pocket — sized AC Line Voltage detectors/testers have a nonconductive outer surface for user safety. Simply touch the tip of the tester to an outlet, terminal strip or cord; if the tip glows red and unit beeps, there's voltage in the line. Ideal for quick checks for energized circuits or ground faults.
Available for sale in the United States and Canada only.
Two year warranty
Specifications
  • DescriptionPocket-Sized Voltage Detectors, 90 to 1000 V AC; 5/PK
  • Warranty2 Years