Fluke TL220 Industrial Test Lead Set
Representative image only.
Fluke TL220 Industrial Test Lead Set
Extech | Mfr: TL220| Item: EW-26002-30
High performance at an economical price
$106.20USD / Set of 2
$118.00USD / Set of 2
$106.20USD / Set of 2
$118.00USD / Set of 2
Buy It Together
Current Item
Fluke TL220 Industrial Test Lead Set
Fluke TL220 Industrial Test Lead Set
2600230
$106.20USD / Set of 2
$118.00USD / Set of 2
+
Fluke TLK-225 SureGrip Master Accessory Kit with Pouch
2000854
$220.50USD / Each
$245.00USD / Each
=
Total Price 326.70 ADD 2 ITEMS TO CART
Product Description
Key Features
  • SureGrip™ design provides a reliable grip
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
Set consists of silicone-insulated TL224 SureGrip test lead set (one red and one black), stainless steel TP220 SureGrip industrial test probe set (one red, one black), and AC220 SureGrip industrial alligator clip set (one red, one black).
1000 V, 10 A maximum.
Shrouded right-angle connectors.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionIndustrial Test Lead Set