Graphic Controls 00983627 Circular Chart; American Meter/Barton, 8 day
Representative image only.
Graphic Controls 00983627 Circular Chart; American Meter/Barton, 8 day
Graphic Controls | Mfr: 00983627| Item: EW-49000-60
Get exacting performance that meets American Gas Association (AGA) standards
$61.39USD / Box of 100
$87.70USD / Box of 100
$61.39USD / Box of 100
$87.70USD / Box of 100
Product Description
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 100%
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.
1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed8 day
  • Chart Diameter (in)12
  • DescriptionCircular Chart; American Meter/Barton, 8 day