Graphic Controls 31232427 Circular Chart, American Meter/Barton, 24 hour
Representative image only.
Graphic Controls 31232427 Circular Chart, American Meter/Barton, 24 hour
Graphic Controls | Mfr: 31232427| Item: EW-49000-55
Get exacting performance that meets American Gas Association (AGA) standards
$61.39USD / Box of 100
$87.70USD / Box of 100
$61.39USD / Box of 100
$87.70USD / Box of 100
Product Description
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 20,000
  • 1 year warranty
More About this Item
Use with American and Barton chart recorders.
1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed1 day
  • Chart Diameter (in)12
  • DescriptionCircular Chart, American Meter/Barton, 24 hour