Hach 14364-01 Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA
Representative image only.
Hach 14364-01 Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA
Hach | Mfr: 1436401| Item: UX-24908-76
$32.60USD / Each
$32.60USD / Each
Buy It Together
Current Item
Hach 14364-01 Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA
Hach 14364-01 Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA
2490876
$32.60USD / Each
+ =
Total Price 378.60 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These compact 13-mm, high-density polyethylene cartridges save storage space, lower shipping costs, and are ideal for fast accurate titrations anywhere. Cartridges maintain titrants at high purity by eliminating evaporation and air space that could lead to deterioration. Cartridges provide 100 typical titrations. Low-volume use minimizes waste and saves money.
Specifications
  • DescriptionTitration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA