Hach Titration Cartridges
These compact 13-mm, high-density polyethylene cartridges save storage space, lower shipping costs, and are ideal for fast accurate titrations anywhere. Cartridges maintain titrants at high purity by eliminating evaporation and air space that could lead to deterioration. Cartridges provide 100 typical titrations. Low-volume use minimizes waste and saves money.
$32.60 - $41.30USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach 14379-01 Titration Cartridge, Acid/Base, 1-10 mg/L Acid, 1.6N NaOH EW-24908-60 Mfr # 1437901
COMPARE
Titration Cartridge, Acid/Base, 1-10 mg/L Acid, 1.6N NaOH
$38.90USD / Each
Hach 14388-01 Titration Cartridge, Alkalinity, 10-160 mg/L CaCO3, 0.16N H2SO4 EW-24908-64 Mfr # 1438801
COMPARE
Titration Cartridge, Alkalinity, 10-160 mg/L CaCO3, 0.16N H2SO4
$33.80USD / Each
Hach 14389-01 Titration Cartridge, Alkalinity, 10-4000 mg/L CaCO3, 1.6N H2SO4 EW-24908-66 Mfr # 1438901
COMPARE
Titration Cartridge, Alkalinity, 10-4000 mg/L CaCO3, 1.6N H2SO4
$34.85USD / Each
Hach 14397-01 Titration Cartridge, Chloride, 100 to 10,000 mg/L; 1.128N AgNO3 EW-24908-68 Mfr # 1439701
COMPARE
Titration Cartridge, Chloride, 100 to 10,000 mg/L; 1.128N AgNO3
$40.10USD / Each
Hach 14396-01 Titration Cartridge, Chloride, 10 to 100 mg/L; 1.128N AgNO3 EW-24908-72 Mfr # 1439601
COMPARE
Titration Cartridge, Chloride, 10 to 100 mg/L; 1.128N AgNO3
$41.30USD / Each
Hach 14364-01 Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA EW-24908-76 Mfr # 1436401
COMPARE
Titration Cartridge, Hardness/Calcium, 10 to 160 mg/L, 0.08M EDTA
$32.60USD / Each
Hach 14399-01 Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA EW-24908-78 Mfr # 1439901
COMPARE
Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA
$33.25USD / Each
Showing 1- 7 of 7