Hach 14399-01 Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA
Representative image only.
Hach 14399-01 Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA
Hach | Mfr: 1439901| Item: EW-24908-78
$33.25USD / Each
$33.25USD / Each
Buy It Together
Current Item
Hach 14399-01 Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA
Hach 14399-01 Titration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA
2490878
$33.25USD / Each
+ =
Total Price 379.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These compact 13-mm, high-density polyethylene cartridges save storage space, lower shipping costs, and are ideal for fast accurate titrations anywhere. Cartridges maintain titrants at high purity by eliminating evaporation and air space that could lead to deterioration. Cartridges provide 100 typical titrations. Low-volume use minimizes waste and saves money.
Specifications
  • DescriptionTitration Cartridge, Hardness/Calcium, 100 to 4000 mg/L, 0.8M EDTA