Hach 22420-00 Colorimeter Test Kit , Aluminum, 0 to 0.8 ppm, 100/Kit

Representative image only.

Hach 22420-00 Colorimeter Test Kit , Aluminum, 0 to 0.8 ppm, 100/Kit

Hach | Mfr: 2242000| Item: EW-99574-03

Environmental test kits for your Hach Colorimeter

$239.00USD / Each

$239.00USD / Each

Buy It Together

Current Item

Hach 22420-00 Colorimeter Test Kit , Aluminum, 0 to 0.8 ppm, 100/Kit
Hach 22420-00 Colorimeter Test Kit , Aluminum, 0 to 0.8 ppm, 100/Kit
9957403

$239.00USD / Each
+ + =
Total Price 1048.85 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Order the test kits for the Hach colorimeter to complete your water testing protocol. All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120 VAC or 99574-65, 240 VAC).

1 Year Warranty
Specifications
  • Test ParametersAluminum
  • Number of Tests Included100
  • Range0 to 0.8 ppm
  • DescriptionColorimeter Test Kit , Aluminum, 0 to 0.8 ppm, 100/Kit
  • Warranty1 year
Reviews
Q&A