Hach 2745125 Free and Total Copper Test Strips, 25
Representative image only.
Hach 2745125 Free and Total Copper Test Strips, 25
Hach | Mfr: 2745125| Item: UX-05546-26
Reduce risk by using as a pre-test validation tool
$42.35USD / Each
$42.35USD / Each
Buy It Together
Current Item
Hach 2745125 Free and Total Copper Test Strips, 25
Hach 2745125 Free and Total Copper Test Strips, 25
0554626
$42.35USD / Each
+ + + + =
Total Price 186.65 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests free and total copper as Cu from 0 to 3 mg/L. 25 test strips included.
ISO9001 Certified Supplier
Specifications
  • TestCopper
  • Measurement Points0, 0.2, 0.5, 1, 3 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionFree and Total Copper Test Strips, 25