Hach PC1R1N Combination pH electrode, 15-ft cable, round glass bulb
Representative image only.
Hach PC1R1N Combination pH electrode, 15-ft cable, round glass bulb
Hach | Mfr: PC1R1N| Item: EW-27400-50
Low Price - High Performance
$475.00USD / Each
$475.00USD / Each
Product Description
Key Features
  • The reference cell double-junction is designed for extended service life and features a built-in solution ground
  • Versatile mounting styles
  • Flat glass electrode features Pt 1000 Ù RTD
  • 3 month warranty
More About this Item
These 3⁄4" combination sensors are designed for specialty applications for immersion or in-line mounting. The reference cell features a double-junction design for extended service life, and a built-in solution ground. The body is molded from chemically-resistant Ryton® PPS, and the reference junction is coaxial porous PTFE. All sensors are rated 0 to 105°C up to 100 psig, and have integral 4.5 m (15 ft.) cables with tinned leads.
Body is constructed of Ryton® PPS with 3⁄4 inch NPT threads on both ends. The sensor can be directly mounted into a standard 3⁄4 inch pipe tee or fastened onto the end of a pipe for immersion mounting. Mounting tees for flow-through mounting are offered in a variety of materials to suit application requirements.
3⁄4 inch combination pH sensors are available with or without a Pt 1000 ohm RTD temperature element.
3 Month Warranty
Specifications
  • Electrode TypeSalt bridge
  • Electrode Body MaterialRyton® PPS
  • Min Temperature (° C)-5
  • Max Temperature (° C)95
  • Max Pressure (PSI)100
  • ConnectorTinned ends
  • DescriptionCombination pH electrode, 15-ft cable, round glass bulb