Hardman 8173A/B-SY Quick-Set Epoxy Syringe, 30 mL; 1/Pack
Representative image only.
Hardman 8173A/B-SY Quick-Set Epoxy Syringe, 30 mL; 1/Pack
Hardman | Mfr: 8173A/B-SY| Item: EW-08778-40
Apply amount necessary, then reseal syringe for later use
$21.00USD / Each
$21.00USD / Each
Product Description
Includes
Syringe cap and ten mixing sticks.
Key Features
  • Quick-dispensing provides convenient repair work
  • 1 year warranty
More About this Item
Double-barrel syringe epoxies provide the appropriate amount of two-component epoxies for the job at hand. There is no need to worry whether or not the correct amount is dispensed. Just dispense, mix and apply to the surface being repaired.
1 Year Warranty
Specifications
  • Curing time (days)1
  • Curing time (mins)1440
  • Curing time (hrs)24
  • Set time (mins)5
  • Min operating temperature (° F)-20
  • Max operating temperature (° F)180
  • DescriptionQuick-Set Epoxy Syringe, 30 mL; 1/Pack