Hardman Double Bubble® Epoxy Pouches, General Purpose, 3.5g; 10/Pk
Representative image only.
Hardman Double Bubble® Epoxy Pouches, General Purpose, 3.5g; 10/Pk
Hardman | Mfr: 4005-BG10| Item: EW-08778-06
Complete selection of epoxies and urethane for all your maintenance needs
$45.40USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$45.40 USD / Pkg of 10
  • 10+$40.86 USD / Pkg of 10
$45.40USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$45.40 USD / Pkg of 10
  • 10+$40.86 USD / Pkg of 10
Product Description
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • A long work-life adhesive for bonding wood, metal, ceramics, concrete, rubber and plastics
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.
Specifications
  • Curing time (days)7
  • Curing time (hrs)96
  • Set time (mins)180
  • Set time (hrs)3
  • Min operating temperature (° F)-30
  • Max operating temperature (° F)200
  • Number of Pieces per Set10
  • DescriptionEpoxy Pouches, General Purpose, 3.5g; 10/Pk