Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk

Representative image only.

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk

Hardy Diagnostics | Mfr: D599| Item: UX-36150-00

The dilution solution saves time and money

$168.50USD / Pkg of 50

$168.50USD / Pkg of 50

Buy It Together

Current Item

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk
3615000

$168.50USD / Pkg of 50
+
GVS Dilution Bottle, Butterfield's Buffer, 99 mL; 72/PK
3615317

$263.50USD / Pkg of 72
=
Total Price 432.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Wide 45-mm opening accommodates large samples
  • Flip-top lid allows for easy one-handed opening

More About this Item

Prefilled dilution vials are designed for the food, dairy, pharmaceutical, and water industries. Eliminate the time and money spent on preparing your own dilution vials. Final pH for all solutions is 7.2 ±0.2 at 25°C. The 145-mL polypropylene vial features a flip-top lid for easy one-handed opening. The peptone water vials are gamma irradiated.

Butterfield's Phosphate Buffer is used as a diluent for the preparation of dilutions in plate count and other laboratory processes. It is recommended as a general diluent in laboratory procedures by the Federal Drug administration and in the Bacteriological Analytical Manual. This product is described in the Standard Methods for the Examination of Water and Wastewater for use in water testing. Contains deionized water and monopotassium phosphate.

Specifications
  • Capacity (mL)99
  • Capacity (oz)3.35
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap ColorNatural
  • Bottle ColorTranslucent
  • Special FeaturesButterfield's Buffer
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)45
  • Neck Inside Diameter (ID) (in)1 3/4
  • DescriptionPrefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk
Reviews
Q&A