Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vials
The dilution solution saves time and money
  • Wide 45-mm opening accommodates large samples
  • Flip-top lid allows for easy one-handed opening
Prefilled dilution vials are designed for the food, dairy, pharmaceutical, and water industries. Eliminate the time and money spent on preparing your own dilution vials. Final pH for all solutions is 7.2 ±0.2 at 25°C. The 145-mL polypropylene vial features a flip-top lid for easy one-handed opening. The peptone water vials are gamma irradiated.
$164.00 - $772.00USD / Pkg of 50
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Special Features Description
Availability Pricing
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk EW-36150-00 Mfr # D599
COMPARE
99 Butterfield's Buffer Prefilled Dilution Vial, Butterfield's Buffer, 99 mL; 50/Pk
$168.50USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Butterfield's Buffer, 90 mL; 50/Pk EW-36150-02 Mfr # D590
COMPARE
90 Butterfield's Buffer Prefilled Dilution Vial, Butterfield's Buffer, 90 mL; 50/Pk
$167.25USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 99 mL; 50/Pk EW-36150-10 Mfr # D699
COMPARE
99 Phosphate Buffer with Magnesium Chloride Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 99 mL; 50/Pk
$169.50USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk EW-36150-12 Mfr # D690
COMPARE
90 Phosphate Buffer with Magnesium Chloride Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk
$164.00USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 99 mL; 50/Pk EW-36150-20 Mfr # D299
COMPARE
99 Peptone Water Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 99 mL; 50/Pk
$612.00USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk EW-36150-22 Mfr # D290
COMPARE
90 Peptone Water Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk
$627.00USD / Pkg of 50
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Buffered Peptone Water, Gamma Irradiated, 99 mL; 50/Pk EW-36150-30 Mfr # D089
COMPARE
99 Buffered Peptone Water Prefilled Dilution Vial, Buffered Peptone Water, Gamma Irradiated, 99 mL; 50/Pk
$772.00USD / Pkg of 50
Showing 1- 7 of 7