Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk

Representative image only.

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk

Hardy Diagnostics | Mfr: D690| Item: EW-36150-12

The dilution solution saves time and money

$164.00USD / Pkg of 50

$164.00USD / Pkg of 50

Buy It Together

Current Item

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk
3615012

$164.00USD / Pkg of 50
+
GVS Dilution Bottle, Butterfield's Buffer, 99 mL; 72/PK
3615317

$263.50USD / Pkg of 72
=
Total Price 427.50 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Wide 45-mm opening accommodates large samples
  • Flip-top lid allows for easy one-handed opening

More About this Item

Prefilled dilution vials are designed for the food, dairy, pharmaceutical, and water industries. Eliminate the time and money spent on preparing your own dilution vials. Final pH for all solutions is 7.2 ±0.2 at 25°C. The 145-mL polypropylene vial features a flip-top lid for easy one-handed opening. The peptone water vials are gamma irradiated.

Phosphate Buffer with Magnesium Chloride is used as a diluent for the preparation of dilutions in plate counts in the dairy and food industries. It is recommended by the APHA for the recovery of injured microorganisms from dairy and food samples. Contains deionized water, monopotassium phosphate, and magnesium chloride.

Specifications
  • Capacity (mL)90
  • Capacity (oz)3.04
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap ColorNatural
  • Bottle ColorTranslucent
  • AutoclavableNo
  • Special FeaturesPhosphate Buffer with Magnesium Chloride
  • Neck Inside Diameter (ID) (mm)45
  • Neck Inside Diameter (ID) (in)1 3/4
  • DescriptionPrefilled Dilution Vial, Phosphate Buffer with Magnesium Chloride, 90 mL; 50/Pk
Reviews
Q&A