Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk

Representative image only.

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk

Hardy Diagnostics | Mfr: D290| Item: EW-36150-22

The dilution solution saves time and money

$627.00USD / Pkg of 50

$627.00USD / Pkg of 50

Buy It Together

Current Item

Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk
Hardy Diagnostics Dilu-Lok™ Prefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk
3615022

$627.00USD / Pkg of 50
+
GVS Dilution Bottle, Butterfield's Buffer, 99 mL; 72/PK
3615317

$263.50USD / Pkg of 72
=
Total Price 890.50 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Wide 45-mm opening accommodates large samples
  • Flip-top lid allows for easy one-handed opening

More About this Item

Prefilled dilution vials are designed for the food, dairy, pharmaceutical, and water industries. Eliminate the time and money spent on preparing your own dilution vials. Final pH for all solutions is 7.2 ±0.2 at 25°C. The 145-mL polypropylene vial features a flip-top lid for easy one-handed opening. The peptone water vials are gamma irradiated.

Peptone Water is used for the cultivation of non-fastidious organisms such as Salmonella, for performing the indole test, and for studying fermentation patterns. Contains deionized water, monopotassium phosphate, and sodium chloride. These bottles are gamma irradiated.

Specifications
  • Capacity (mL)90
  • Capacity (oz)3.04
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap ColorNatural
  • Bottle ColorTranslucent
  • Special FeaturesPeptone Water
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)45
  • Neck Inside Diameter (ID) (in)1 3/4
  • DescriptionPrefilled Dilution Vial, Peptone Water, 0.1%, Gamma Irradiated, 90 mL; 50/Pk
Reviews
Q&A