Honeywell S1299C Uvex Eyewear, Clear Lens, Blue Frame
Representative image only.
Honeywell S1299C Uvex Eyewear, Clear Lens, Blue Frame
Honeywell | Mfr: S1299C| Item: EW-86321-10
Available for sale in the US only
$21.80USD / Each
BUY MORE & SAVE
  • 1-9$21.80 USD / Each
  • 10+$19.62 USD / Each
$21.80USD / Each
BUY MORE & SAVE
  • 1-9$21.80 USD / Each
  • 10+$19.62 USD / Each
Product Description
Key Features
  • A comfortable fit for virtually any size face
  • Adjustable temple length and lens inclination
More About this Item
An anti-fog Uvextreme® coating with additional anti-scratch and anti-static properties.
Compliance: ANSI Z87.1-2010, CSA Z94.3, military V0
Specifications
  • Frame ColorBlue
  • Temple StyleDuoflex®
  • Lens ColorClear
  • Lens CoatingUvextreme® (reduces fogging, scratches, and static)
  • ComplianceANSI Z87.1-2010; CSA Z94.3; military V0
  • DescriptionEyewear, Clear Lens, Blue Frame