ICI Quick Ship Envision Female Quick-Connect Fittings, Chrome-Plated Brass, Vacuum, Yellow; 1/Each
Representative image only.
ICI Quick Ship Envision Female Quick-Connect Fittings, Chrome-Plated Brass, Vacuum, Yellow; 1/Each
ICI Quick Ship | Mfr: CPQCFL-B-3M-K4| Item: UX-78984-45
Quick connect fittings designed specifically for Envision workstation plumbing fixtures
$287.75USD / Each
$287.75USD / Each
Product Description
Key Features
  • Conveniently color labeled for easy coupling
  • Available for air, vacuum, and nitrogen services
  • Include all hardware necessary to attach to the service panel
  • Constructed of durable chrome-plated brass
  • Compatible with plumbling assemblies 78984-39 and 78984-42
  • 2 year warranty
More About this Item
ICI Quick Ship female quick-disconnect fittings are designed to mate with the Envision plumbing assemblies. Simply mount with the overhead service panel and connect it from the workstation to the building services. Fittings are designed to only couple with the same key, which is conveniently color coded in orange, yellow, and brown. All fittings include 3/8 in stems and hardware to attach to the overhead service panel.
Two year warranty
Specifications
  • Fitting MaterialChrome-Plated Brass
  • Chemical ResistanceFor Vacuum
  • ColorYellow
  • DescriptionEnvision Female Quick-Connect Fittings, Vacuum, Yellow
  • Warranty2 Years