ICI Quick Ship Envision Female Quick Connect Fittings, Chrome-Plated Brass
Quick connect fittings designed specifically for Envision workstation plumbing fixtures
  • Conveniently color labeled for easy coupling
  • Available for air, vacuum, and nitrogen services
  • Include all hardware necessary to attach to the service panel
  • Constructed of durable chrome-plated brass
ICI Quick Ship female quick-disconnect fittings are designed to mate with the Envision plumbing assemblies. Simply mount with the overhead service panel and connect it from the workstation to the building services. Fittings are designed to only couple with the same key, which is conveniently color coded in orange, yellow, and brown. All fittings include 3/8 in stems and hardware to attach to the overhead service panel.
$253.25 - $287.75USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chemical Resistance Color Description
Availability Pricing
ICI Quick Ship Envision Female Quick-Connect Fittings, Chrome-Plated Brass, Air, Orange; 1/Each EW-78984-44 Mfr # CPQCFL-B-3M-K2
COMPARE
For Air Orange Envision Female Quick-Connect Fittings, Air, Orange
$287.75USD / Each
ICI Quick Ship Envision Female Quick-Connect Fittings, Chrome-Plated Brass, Vacuum, Yellow; 1/Each EW-78984-45 Mfr # CPQCFL-B-3M-K4
COMPARE
For Vacuum Yellow Envision Female Quick-Connect Fittings, Vacuum, Yellow
$287.75USD / Each
ICI Quick Ship Envision Female Quick-Connect Fittings, Chrome-Plated Brass, Nitrogen, Brown; 1/Each EW-78984-46 Mfr # CPQCFL-B-3M-K8
COMPARE
For Nitrogen Brown Envision Female Quick-Connect Fittings, Nitrogen, Brown
$253.25USD / Each