ICI Quick Ship Fan Blower Switch, 1 hp
Representative image only.
ICI Quick Ship Fan Blower Switch, 1 hp
ICI Quick Ship | Mfr: CP2510FS1P| Item: EW-78981-03
Enhance your fume hood performance
$777.00USD / Each
$777.00USD / Each
Product Description
Key Features
  • Ideal for use with 1 hp fan
  • 2 year warranty
More About this Item
Choose from a wide range of accessories to enhance the performance of your fume hood.
The manual fan motor starter switch is a toggle style, single pole, with red pilot light. It features a stainless steel cover plate and one thermal unit.
For sale in U.S. only!
Two year warranty
Specifications
  • Compatible WithICI Quick Ship ducted fume hoods
  • DescriptionICI Quick Ship Fan Blower Switch, 1 hp
  • Warranty2 Years