ICI Quick Ship Foot Operated Faucet with Vacuum Breaker
Representative image only.
ICI Quick Ship Foot Operated Faucet with Vacuum Breaker
ICI Quick Ship | Mfr: CPL084-8-3001-BH| Item: UX-78984-76
Faucets for a quick delivery furniture program that builds complete labs
$834.00USD / Each
$834.00USD / Each
Product Description
Key Features
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
  • Order optional aerator separately
  • 2 year warranty
More About this Item
Choose from mixing faucets, cold water faucets, or a specialty faucets such as the foot-operated and DI/RO faucets.
Mixing faucets and cold water faucets are deck mounted with an option for a vacuum breaker. The material is chrome-plated brass. They feature an 8” swivel gooseneck with four arm or blade-handles. Valves are self-contained and have replaceable stainless-steel seats. The foot operated faucet has all the features of the mixing and cold water faucets plus a floor-mounted foot valve.
The DI/RO faucet is deck mounted for distilled, deionized, or reverse osmosis water. It has a 6” spread that features exterior brass casing with interior polypropylene lining. The valve is a diaphragm-type and the handle is a nylon toggle-style lever with combination manual/self-closing.
Two year warranty
Specifications
  • DescriptionFoot Operated Faucet with Vacuum Breaker
  • Warranty2 Years