ICI Quick Ship Faucets
Faucets for a quick delivery furniture program that builds complete labs
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
  • Order optional aerator separately
Choose from mixing faucets, cold water faucets, or a specialty faucets such as the foot-operated and DI/RO faucets.
Mixing faucets and cold water faucets are deck mounted with an option for a vacuum breaker. The material is chrome-plated brass. They feature an 8” swivel gooseneck with four arm or blade-handles. Valves are self-contained and have replaceable stainless-steel seats. The foot operated faucet has all the features of the mixing and cold water faucets plus a floor-mounted foot valve.
The DI/RO faucet is deck mounted for distilled, deionized, or reverse osmosis water. It has a 6” spread that features exterior brass casing with interior polypropylene lining. The valve is a diaphragm-type and the handle is a nylon toggle-style lever with combination manual/self-closing.
$35.30 - $834.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
ICI Quick Ship Mixing Faucet, Four-arm Handle, with Vacuum Breaker EW-78984-68 Mfr # CPL412-8VB
COMPARE
Mixing Faucet, Four-arm Handle, with Vacuum Breaker
$619.00USD / Each
ICI Quick Ship Mixing Faucet, Four-arm Handle, without Vacuum Breaker EW-78984-69 Mfr # CPL412-8
COMPARE
Mixing Faucet, Four-arm Handle, without Vacuum Breaker
$462.00USD / Each
ICI Quick Ship Mixing Faucet, Blade Handle, with Vacuum Breaker EW-78984-70 Mfr # CPL412-8VB-BH
COMPARE
Mixing Faucet, Blade Handle, with Vacuum Breaker
$661.00USD / Each
ICI Quick Ship Mixing Faucet, Blade Handle, without Vacuum Breaker EW-78984-71 Mfr # CPL412-8-BH
COMPARE
Mixing Faucet, Blade Handle, without Vacuum Breaker
$503.00USD / Each
ICI Quick Ship Cold Water Faucet, Four-arm Handle, with Vacuum Breaker EW-78984-72 Mfr # CPL611-8VB
COMPARE
Cold Water Faucet, Four-arm Handle, with Vacuum Breaker
$464.50USD / Each
ICI Quick Ship Cold Water Faucet, Four-arm Handle, without Vacuum Breaker EW-78984-73 Mfr # CPL611-8
COMPARE
Cold Water Faucet, Four-arm Handle, without Vacuum Breaker
$275.50USD / Each
ICI Quick Ship Cold Water Faucet, Blade Handle, with Vacuum Breaker EW-78984-74 Mfr # CPL611-8VB-BH
COMPARE
Cold Water Faucet, Blade Handle, with Vacuum Breaker
$515.00USD / Each
ICI Quick Ship Cold Water Faucet, Blade Handle, without Vacuum Breaker EW-78984-75 Mfr # CPL611-8-BH
COMPARE
Cold Water Faucet, Blade Handle, without Vacuum Breaker
$295.75USD / Each
ICI Quick Ship Foot Operated Faucet with Vacuum Breaker EW-78984-76 Mfr # CPL084-8-3001-BH
COMPARE
Foot Operated Faucet with Vacuum Breaker
$834.00USD / Each
ICI Quick Ship DI/RO Faucet EW-78984-77 Mfr # CPL7833MSC
COMPARE
DI/RO Faucet
$782.00USD / Each
ICI Quick Ship Aerator EW-78984-78 Mfr # CPBO055
COMPARE
Aerator
$35.30USD / Each
Showing 1- 11 of 11