ICI Quick Ship Fume Hood Alarm
Representative image only.
ICI Quick Ship Fume Hood Alarm
ICI Quick Ship | Mfr: CPAFA4000| Item: EW-78981-02
Enhance your fume hood performance
$1,810.00USD / Each
$1,810.00USD / Each
Product Description
Key Features
  • Each fume hoood is pre-punched for fast, easy installation of alarm
  • Provides both visual and audiable alarms
  • Shipped loose and installed on site
  • 2 year warranty
More About this Item
Choose from a wide range of accessories to enhance the performance of your fume hood.
The alarm moitor provides a digital readout of fume hood face velocity and sounds an alarm when airflow falls below safe levels. It contains test/reset button allowing the user to verify alarm readiness. The thermostat sensor is mounted through the side wall of the fume hood while the sensor unit is mounted on the fascia.
For sale in U.S. only!
Two year warranty
Specifications
  • Compatible WithICI Quick Ship ducted fume hoods
  • DescriptionICI Quick Ship Fume Hood Alarm
  • Warranty2 Years